Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUP35
Entrez Gene ID
129401
Gene Name
nucleoporin 35
Gene Aliases
MP-44, MP44, NP44, NUP53
Chromosome Location
Chr.2: 183117490 - 183161684 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_109856.1 4 - 5 501 75
GenBank mRNA BI821231.1 - 4 - 5 437 75


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BU853804.1 - 4 - 5 465 Chr. 2 - 183117520 - 183159560 [+] on Build GRCh38 75
GenBankmRNA BF970614.1 - 4 - 5 436 Chr. 2 - 183124431 - 183159528 [+] on Build GRCh38 75


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream dual specificity phosphatase 19 DUSP19 DUSP17, LMWDSP3, SKRP1, TS-DSP1 -33.6kb Sense Chr. - 183078559 - 183100008 on Build GRCh38
Downstream microRNA 548ae-1 MIR548AE1 1.2mb Sense Chr. - 184378975 - 184379044 on Build GRCh38

View Full Product Details