Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- BEAN1-AS1
- Entrez Gene ID
-
101927726
- Gene Name
- BEAN1 antisense RNA 1
- Gene Aliases
-
CTD-2258A20.5
- Chromosome Location
-
Chr.16: 66469796 - 66476402 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_109960.1 |
|
1 - 2 |
141 |
142 |
GenBank mRNA |
DA862085.1 |
- |
1 - 2 |
141 |
142 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
long intergenic non-protein coding RNA 920 |
LINC00920 |
|
-64.7kb
|
Anti-sense |
Chr. - 66408524 - 66410901 on Build GRCh38 |
Downstream |
thymidine kinase 2, mitochondrial |
TK2 |
MTDPS2, MTTK, SCA31 |
31.6kb
|
Sense |
Chr. - 66508003 - 66550412 on Build GRCh38 |
View Full Product Details