Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LOC101927446
- Entrez Gene ID
-
101927446
- Gene Name
- uncharacterized LOC101927446
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 90335223 - 90344094 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_110085.1 |
|
4 - 5 |
733 |
120 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
cilia and flagella associated protein 69 |
CFAP69 |
C7orf63, FAP69 |
-16.2kb
|
Sense |
Chr. - 90245174 - 90320964 on Build GRCh38 |
Downstream |
GTP binding protein 10 |
GTPBP10 |
ObgH2, UG0751c10 |
5.1kb
|
Sense |
Chr. - 90346665 - 90391455 on Build GRCh38 |
View Full Product Details