Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LINC01284
- Entrez Gene ID
-
101926971
- Gene Name
- long intergenic non-protein coding RNA 1284
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 51095836 - 51117830 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_110382.1 |
|
4 - 5 |
234 |
71 |
GenBank mRNA |
BX112678.1 |
- |
4 - 5 |
237 |
71 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB302181.1 |
- |
5 - 6 |
|
|
Chr. X - 51095837 - 51101600 [+] on Build GRCh38 |
71 |
GenBankmRNA |
H21441.1 |
- |
4 - 5 |
239 |
|
Chr. X - 51101267 - 51171391 [-] on Build GRCh38 |
71 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
bone morphogenetic protein 15 |
BMP15 |
GDF9B, ODG2, POF4 |
-184.8kb
|
Anti-sense |
Chr. - 50910735 - 50916641 on Build GRCh38 |
Downstream |
nudix hydrolase 10 |
NUDT10 |
APS2, DIPP3-alpha, DIPP3a |
230.6kb
|
Anti-sense |
Chr. - 51332231 - 51337525 on Build GRCh38 |
View Full Product Details