Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LINC01284
- Entrez Gene ID
-
101926971
- Gene Name
- long intergenic non-protein coding RNA 1284
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 51095836 - 51117830 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_110382.1 |
|
8 - 9 |
583 |
92 |
GenBank mRNA |
BG212281.1 |
- |
1 - 2 |
158 |
92 |
BX112678.1 |
- |
8 - 9 |
586 |
92 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG205519.1 |
- |
1 - 2 |
158 |
|
Chr. X - 51095836 - 51098053 [-] on Build GRCh38 |
92 |
GenBankmRNA |
DB302181.1 |
- |
1 - 2 |
|
|
Chr. X - 51095837 - 51101600 [+] on Build GRCh38 |
94 |
GenBankmRNA |
H22369.1 |
- |
1 - 2 |
|
|
Chr. X - 51095838 - 51101134 [+] on Build GRCh38 |
92 |
GenBankmRNA |
BG200913.1 |
- |
1 - 2 |
|
|
Chr. X - 51095848 - 51101616 [+] on Build GRCh38 |
92 |
GenBankmRNA |
BG214407.1 |
- |
1 - 2 |
156 |
|
Chr. X - 51095846 - 51098043 [-] on Build GRCh38 |
92 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
bone morphogenetic protein 15 |
BMP15 |
GDF9B, ODG2, POF4 |
-179.2kb
|
Anti-sense |
Chr. - 50910735 - 50916641 on Build GRCh38 |
Downstream |
nudix hydrolase 10 |
NUDT10 |
APS2, DIPP3-alpha, DIPP3a |
234.2kb
|
Anti-sense |
Chr. - 51332231 - 51337525 on Build GRCh38 |
View Full Product Details