Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LOC101929717
- Entrez Gene ID
-
101929717
- Gene Name
- uncharacterized LOC101929717
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 113142350 - 113167731 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_110823.1 |
|
2 - 3 |
210 |
73 |
GenBank mRNA |
AK096271.1 |
- |
2 - 3 |
305 |
73 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DA697791.1 |
- |
2 - 3 |
305 |
|
Chr. 3 - 113144284 - 113167819 [-] on Build GRCh38 |
73 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
nucleolus and neural progenitor protein |
NEPRO |
C3orf17, NET17 |
-133.4kb
|
Sense |
Chr. - 113002444 - 113019733 on Build GRCh38 |
Downstream |
BOC cell adhesion associated, oncogene regulated |
BOC |
CDON2 |
45.8kb
|
Anti-sense |
Chr. - 113211003 - 113287463 on Build GRCh38 |
View Full Product Details