The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_111936.1 | 1 - 2 | 108 | 95 | |
GenBank mRNA | DA085292.1 | - | 1 - 2 | 108 | 95 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | G protein-coupled receptor 89B | GPR89B | GPHR, GPR89, GPR89C, SH120, UNQ192 | -16.9kb | Anti-sense | Chr. - 147928393 - 147993523 on Build GRCh38 |
Downstream | RNA, variant U1 small nuclear 7 | RNVU1-7 | RNU1-26P, RNU1-6, RNU1-6P, RNU1-9, RNU1-9P, RNVU1-9, vU1.7, vU1.9 | 23.8kb | Sense | Chr. - 148038753 - 148038916 on Build GRCh38 |