Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- DUSP10
- Entrez Gene ID
-
11221
- Gene Name
- dual specificity phosphatase 10
- Gene Aliases
-
MKP-5, MKP5
- Chromosome Location
-
Chr.1: 221701420 - 221742176 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_111939.1 |
|
1 - 2 |
57 |
85 |
GenBank mRNA |
BC020608.1 |
- |
1 - 2 |
57 |
85 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC400804 |
C1orf140 |
|
-369.8kb
|
Sense |
Chr. - 221329928 - 221336296 on Build GRCh38 |
Downstream |
uncharacterized LOC101929771 |
LOC101929771 |
|
90.8kb
|
Sense |
Chr. - 221827666 - 221840666 on Build GRCh38 |
View Full Product Details