Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- AADACL3
- Entrez Gene ID
-
126767
- Gene Name
- arylacetamide deacetylase like 3
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 12716115 - 12728759 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_111984.1 |
|
1 - 2 |
225 |
75 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
arylacetamide deacetylase like 4 |
AADACL4 |
|
-49.0kb
|
Sense |
Chr. - 12644547 - 12667086 on Build GRCh38 |
Downstream |
chromosome 1 open reading frame 158 |
C1orf158 |
|
25.2kb
|
Sense |
Chr. - 12746186 - 12761169 on Build GRCh38 |
View Full Product Details