Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SLC26A5
- Entrez Gene ID
-
375611
- Gene Name
- solute carrier family 26 member 5
- Gene Aliases
-
DFNB61, PRES
- Chromosome Location
-
Chr.7: 103352598 - 103446177 on Build GRCh38
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
proteasome 26S subunit, ATPase 2 |
PSMC2 |
MSS1, Nbla10058, S7 |
-9.1kb
|
Anti-sense |
Chr. - 103344405 - 103369395 on Build GRCh38 |
Downstream |
uncharacterized LOC101927870 |
LOC101927870 |
|
65.9kb
|
Anti-sense |
Chr. - 103445207 - 103514007 on Build GRCh38 |
View Full Product Details