Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LOC102724094
- Entrez Gene ID
-
102724094
- Gene Name
- uncharacterized LOC102724094
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 101014320 - 101017608 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_120520.1 |
|
1 - 2 |
195 |
108 |
GenBank mRNA |
AA025193.1 |
- |
1 - 2 |
170 |
108 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AA729888.1 |
- |
2 - 3 |
|
|
Chr. 7 - 101014322 - 101017576 [+] on Build GRCh38 |
108 |
GenBankmRNA |
AI382026.1 |
- |
2 - 3 |
|
|
Chr. 7 - 101014321 - 101017563 [+] on Build GRCh38 |
108 |
GenBankmRNA |
AI223808.1 |
- |
2 - 3 |
|
|
Chr. 7 - 101014325 - 101017601 [+] on Build GRCh38 |
108 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
mucin 12, cell surface associated |
MUC12 |
MUC-11, MUC-12, MUC11 |
|
Anti-sense |
Chr. - 100969623 - 101018949 on Build GRCh38 |
Downstream |
mucin 17, cell surface associated |
MUC17 |
MUC-17, MUC-3, MUC3 |
2.5kb
|
Anti-sense |
Chr. - 101020083 - 101058859 on Build GRCh38 |
View Full Product Details