Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PGM5-AS1
- Entrez Gene ID
-
572558
- Gene Name
- PGM5 antisense RNA 1
- Gene Aliases
-
FAM233A
- Chromosome Location
-
Chr.9: 68355164 - 68357866 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_121191.1 |
|
1 - 2 |
262 |
128 |
GenBank mRNA |
BC017938.1 |
- |
1 - 2 |
262 |
128 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
forkhead box D4-like 3 |
FOXD4L3 |
FOXD4L2, FOXD6 |
-50.1kb
|
Anti-sense |
Chr. - 68302867 - 68305084 on Build GRCh38 |
Downstream |
transmembrane protein 252 |
TMEM252 |
C9orf71 |
172.8kb
|
Sense |
Chr. - 68536580 - 68540867 on Build GRCh38 |
View Full Product Details