Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CDC37L1-AS1
- Entrez Gene ID
-
101929351
- Gene Name
- CDC37L1 antisense RNA 1 (head to head)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 4676600 - 4679480 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_121597.1 |
|
1 - 2 |
173 |
53 |
GenBank mRNA |
CR737367.1 |
- |
1 - 2 |
172 |
53 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AI702492.1 |
- |
1 - 2 |
|
|
Chr. 9 - 4676600 - 4679450 [+] on Build GRCh38 |
53 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
spermatogenesis associated 6 like |
SPATA6L |
C9orf68, bA6J24.2 |
-9.8kb
|
Sense |
Chr. - 4540092 - 4666674 on Build GRCh38 |
Downstream |
cell division cycle 37 like 1 |
CDC37L1 |
CDC37B, HARC |
73.0
|
Anti-sense |
Chr. - 4679553 - 4708399 on Build GRCh38 |
View Full Product Details