Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LINC01291
- Entrez Gene ID
-
102724515
- Gene Name
- long intergenic non-protein coding RNA 1291
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 74909131 - 74938418 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_125792.1 |
|
2 - 3 |
165 |
92 |
GenBank mRNA |
CV382059.1 |
- |
2 - 3 |
186 |
92 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105374809 |
LOC105374809 |
|
|
Anti-sense |
Chr. - 74925566 - 74958892 on Build GRCh38 |
Downstream |
long intergenic non-protein coding RNA 1293 |
LINC01293 |
|
7.1kb
|
Sense |
Chr. - 74939347 - 74942670 on Build GRCh38 |
View Full Product Details