Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RPS18P9
- Entrez Gene ID
-
645958
- Gene Name
- ribosomal protein S18 pseudogene 9
- Gene Aliases
-
RPS18_6_767
- Chromosome Location
-
Chr.6: 149593616 - 149594602 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_077247.1 |
|
1 - 1 |
420 |
75 |
GenBank mRNA |
EB387363.1 |
- |
1 - 1 |
420 |
75 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
glycoprotein integral membrane 1 |
GINM1 |
C6orf72, dJ12G14.2 |
-2.7kb
|
Sense |
Chr. - 149566392 - 149590931 on Build GRCh38 |
Downstream |
katanin catalytic subunit A1 |
KATNA1 |
|
271.0
|
Anti-sense |
Chr. - 149594873 - 149649018 on Build GRCh38 |
View Full Product Details