Product Details

Assay Reference Genome
Location

Chr.X:41471955 on build GRCh38
Cytoband
Xp11.4
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
60506
Gene Symbol
NYX
Gene Name
nyctalopin
Gene Aliases
CLRP, CSNB1, CSNB1A, CSNB4, NBM1
Location
Chr.X:41447460-41476414 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
NYX NM_022567.2 NP_072089.1
XM_017029709.1 XP_016885198.1
AF254868.1 AAG42685.1
AJ278865.1 CAC19014.1
BC112242.1 AAI12243.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv508763 ChrX:41415959 - 41531609 on Build GRCh38 Insertion CASK NYX
esv33720 ChrX:40891487 - 43157518 on Build GRCh38 Loss LOC105373185 CASK GPR34 DDX3X PPP1R2P9 GPR82 LOC107985678 LOC643043 USP9X NYX

View Full Product Details