Product Details

Assay Reference Genome
Location

Chr.3:87234539 on build GRCh38
Cytoband
3p11.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
25978
Gene Symbol
CHMP2B
Gene Name
charged multivesicular body protein 2B
Gene Aliases
ALS17, CHMP2.5, DMT1, VPS2-2, VPS2B
Location
Chr.3:87227263-87255548 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
CHMP2B NM_001244644.1 NP_001231573.1
NM_014043.3 NP_054762.2
AF151842.1 AAD34079.1
AK002165.1
AK002180.1
AK222654.1 BAD96374.1
AK296072.1
AK316585.1
AL080122.1 CAB45721.1
BC001553.1 AAH01553.1
BI821848.1
BM828410.1
CR533456.1 CAG38487.1
DA426159.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv953231 Chr3:87227951 - 87252550 on Build GRCh38 Duplication CHMP2B
esv3568885 Chr3:87182571 - 87518398 on Build GRCh38 Loss POU1F1 MIR4795 CHMP2B
nsv527279 Chr3:86796784 - 87814130 on Build GRCh38 Gain POU1F1 LINC00506 MIR4795 HTR1F VGLL3 CHMP2B

View Full Product Details