Target Copy Number Variation Details
- DGV Version
- Release date: 2016-05-15, GRCh GRCh38
Target Variation ID |
Location |
CNV Subtype |
Genes |
nsv596369 |
Chr4:184907523 - 185314394 on Build GRCh38 |
Loss |
LOC101928877
MIR4455
SLC25A4
SNX25
HELT
CFAP97
|
Product Details
- Assay Reference Genome
Location
Chr.4:185147450 on build GRCh38
- Cytoband
- 4q35.1
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 291
- Gene Symbol
- SLC25A4
- Gene Name
- solute carrier family 25 member 4
- Gene Aliases
- AAC1, ANT, ANT 1, ANT1, MTDPS12, PEO2, PEO3, PEOA2, T1
- Location
- Chr.4:185143263-185150384 on build GRCh38
- Assay Gene Location
- Within Exon 4
Gene Symbol |
Transcript Accession |
Exon Location |
Assay Location |
Protein ID |
SLC25A4 |
NM_001151.3 |
4
|
1486 |
NP_001142.2 |
Target Copy Number Variation Details
- DGV Version
- Release date: 2016-05-15, GRCh GRCh38
Target Variation |
Location |
CNV Subtype |
Genes |
nsv596369 |
Chr4:184907523 - 185314394 on Build GRCh38 |
Loss |
LOC101928877
MIR4455
SLC25A4
SNX25
HELT
CFAP97
|
View Full Product Details