The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199641.1 | NP_001186570.1 | 2 - 3 | 164 | 103 |
GenBank mRNA | AK295908.1 | - | 2 - 3 | 145 | 103 |