Product Details

Assay Reference Genome
Location

Chr.7:101050394 on build GRCh38
Cytoband
7q22.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
140453
Gene Symbol
MUC17
Gene Name
mucin 17, cell surface associated
Gene Aliases
MUC-17, MUC-3, MUC3
Location
Chr.7:101020083-101058859 on build GRCh38
Assay Gene Location
Within Intron 6
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
MUC17 NM_001040105.1 NP_001035194.1
NR_133665.1
AF430017.1 AAL89737.1
AJ606307.1 CAE54435.1
AJ606308.1 CAE54436.1
AK026404.1
BC126315.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv5873 Chr7:101040374 - 101074468 on Build GRCh38 Insertion MUC17
nsv428181 Chr7:100962999 - 101105965 on Build GRCh38 Gain+Loss MUC17 MUC12 MUC3A TRIM56 LOC102724094
esv2759547 Chr7:100757867 - 101105965 on Build GRCh38 Gain+Loss MUC17 MUC3A SLC12A9 MIR6875 UFSP1 ZAN LOC102724094 MUC12 SRRT EPHB4 TRIM56 ACHE TRIP6
esv2734888 Chr7:99975623 - 101290285 on Build GRCh38 Deletion STAG3L5P-PVRIG2P-PILRB MIR4658 C7orf61 MUC3A COPS6 ZNF3 MIR4653 ACTL6B ZKSCAN1 MIR6840 PMS2P1 ZCWPW1 STAG3 POP7 ZSCAN21 EPHB4 MEPCE MIR106B MBLAC1 CLDN15 MCM7 MIR6875 CNPY4 TFR2 FIS1 NYAP1 MUC12 AZGP1P1 AZGP1 SPDYE3 PILRA NAT16 LOC105375429 AGFG2 PVRIG PCOLCE GATS TSC22D4 PVRIG2P GPC2 MIR25 C7orf43 SRRT SAP25 AP1S1 AP4M1 FBXO24 TRIP6 MUC17 MOSPD3 SLC12A9 UFSP1 STAG3L5P PCOLCE-AS1 PILRB ZAN LRCH4 ZASP LOC102724094 GNB2 LAMTOR4 GAL3ST4 MIR93 MOGAT3 PLOD3 TAF6 VGF SERPINE1 EPO TRIM56 PPP1R35 ZNHIT1 GIGYF1 ACHE
nsv831075 Chr7:101049016 - 101273737 on Build GRCh38 Loss MUC17 CLDN15 MOGAT3 PLOD3 VGF SERPINE1 TRIM56 AP1S1 NAT16 FIS1 ZNHIT1 MIR4653
nsv1033081 Chr7:101026348 - 101128543 on Build GRCh38 Gain MUC17 SERPINE1 TRIM56
nsv831074 Chr7:100963006 - 101102251 on Build GRCh38 Loss MUC17 MUC12 MUC3A TRIM56 LOC102724094

View Full Product Details