Product Details

Assay Reference Genome
Location

Chr.7:101020617 on build GRCh38
Cytoband
7q22.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
140453
Gene Symbol
MUC17
Gene Name
mucin 17, cell surface associated
Gene Aliases
MUC-17, MUC-3, MUC3
Location
Chr.7:101020083-101058859 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
MUC17 NM_001040105.1 NP_001035194.1
NR_133665.1
AJ606307.1 CAE54435.1
AJ606308.1 CAE54436.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv428181 Chr7:100962999 - 101105965 on Build GRCh38 Gain+Loss LOC102724094 MUC12 MUC17 MUC3A TRIM56
esv2759547 Chr7:100757867 - 101105965 on Build GRCh38 Gain+Loss LOC102724094 TRIP6 SRRT ZAN MUC12 EPHB4 ACHE MUC3A UFSP1 MIR6875 SLC12A9 MUC17 TRIM56
esv2734888 Chr7:99975623 - 101290285 on Build GRCh38 Deletion MIR6840 TSC22D4 SPDYE3 TRIP6 SRRT EPHB4 MIR25 PCOLCE-AS1 CNPY4 COPS6 POP7 MCM7 GAL3ST4 PVRIG2P PLOD3 AZGP1 PVRIG NAT16 ZSCAN21 AZGP1P1 FBXO24 AP4M1 MUC3A STAG3L5P-PVRIG2P-PILRB GATS GPC2 PPP1R35 LOC105375429 STAG3L5P MOSPD3 FIS1 NYAP1 SLC12A9 MUC17 PMS2P1 MOGAT3 TFR2 MIR106B ACTL6B VGF MIR4658 ZNHIT1 ZKSCAN1 ZAN MUC12 MBLAC1 MEPCE ZASP TAF6 UFSP1 PCOLCE LAMTOR4 AGFG2 MIR93 STAG3 MIR4653 GIGYF1 ZNF3 SAP25 TRIM56 LOC102724094 AP1S1 ACHE GNB2 MIR6875 PILRA PILRB LRCH4 CLDN15 C7orf61 EPO C7orf43 ZCWPW1 SERPINE1
esv3614313 Chr7:100972001 - 101031293 on Build GRCh38 Gain LOC102724094 MUC12 MUC17
nsv5872 Chr7:100999861 - 101030935 on Build GRCh38 Insertion LOC102724094 MUC12 MUC17
nsv831074 Chr7:100963006 - 101102251 on Build GRCh38 Loss LOC102724094 MUC12 MUC17 MUC3A TRIM56

View Full Product Details