Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
EGFR | NM_005228.3 | NP_005219.2 | ||
NM_201282.1 | NP_958439.1 | |||
NM_201283.1 | NP_958440.1 | |||
NM_201284.1 | NP_958441.1 | |||
AB209442.1 | BAD92679.1 | |||
AF125253.1 | AAG43240.1 | |||
AK225422.1 | ||||
AK290352.1 | ||||
AW163038.1 | ||||
AY550966.1 | AAT52212.1 | |||
AY698023.1 | AAT97978.1 | |||
AY698024.1 | AAT97979.1 | |||
BC094761.1 | AAH94761.1 | |||
DQ088980.1 | ||||
GU255993.1 | ||||
HQ912715.1 | ||||
K03193.1 | AAA52371.1 | |||
U48722.1 | AAC50802.1 | |||
U95089.1 | AAB53063.1 | |||
X00588.1 | CAA25240.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2761332 | Chr7:54574291 - 55097556 on Build GRCh38 | Gain | EGFR SEC61G LOC107986797 LOC100996654 |
nsv607029 | Chr7:54579095 - 55086688 on Build GRCh38 | Gain | EGFR SEC61G LOC107986797 LOC100996654 |
esv2734425 | Chr7:53868335 - 55917254 on Build GRCh38 | Deletion | HPVC1 ELDR PSPHP1 LOC101060341 SEC61G FKBP9P1 VSTM2A-OT1 VOPP1 EGFR SEPT14 VSTM2A EGFR-AS1 LOC107986797 ZNF713 LOC100996654 LINC01445 LANCL2 |
nsv1030676 | Chr7:54583600 - 55151489 on Build GRCh38 | Gain | EGFR SEC61G LOC107986797 LOC100996654 |
nsv5752 | Chr7:54995331 - 55040249 on Build GRCh38 | Deletion | EGFR |