Product Details

Assay Reference Genome
Location

Chr.7:55056179 on build GRCh38
Cytoband
7p11.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
1956
Gene Symbol
EGFR
Gene Name
epidermal growth factor receptor
Gene Aliases
ERBB, ERBB1, HER1, NISBD2, PIG61, mENA
Location
Chr.7:55019032-55207338 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
EGFR NM_005228.3 NP_005219.2
NM_201282.1 NP_958439.1
NM_201283.1 NP_958440.1
NM_201284.1 NP_958441.1
AB209442.1 BAD92679.1
AF125253.1 AAG43240.1
AK225422.1
AK290352.1
AW163038.1
AY550966.1 AAT52212.1
AY698023.1 AAT97978.1
AY698024.1 AAT97979.1
BC094761.1 AAH94761.1
DQ088980.1
GU255993.1
HQ912715.1
K03193.1 AAA52371.1
U48722.1 AAC50802.1
U95089.1 AAB53063.1
X00588.1 CAA25240.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2761332 Chr7:54574291 - 55097556 on Build GRCh38 Gain LOC107986797 SEC61G LOC100996654 EGFR
nsv607029 Chr7:54579095 - 55086688 on Build GRCh38 Gain LOC107986797 SEC61G LOC100996654 EGFR
nsv5753 Chr7:55045323 - 55070844 on Build GRCh38 Insertion EGFR
esv2734425 Chr7:53868335 - 55917254 on Build GRCh38 Deletion SEPT14 SEC61G VOPP1 EGFR HPVC1 LOC101060341 LOC107986797 VSTM2A-OT1 ELDR FKBP9P1 LOC100996654 PSPHP1 EGFR-AS1 VSTM2A LINC01445 LANCL2 ZNF713
nsv1030676 Chr7:54583600 - 55151489 on Build GRCh38 Gain LOC107986797 SEC61G LOC100996654 EGFR

View Full Product Details