Product Details

Assay Reference Genome
Location

Chr.15:38280355 on build GRCh38
Cytoband
15q14
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
161742
Gene Symbol
SPRED1
Gene Name
sprouty related EVH1 domain containing 1
Gene Aliases
NFLS, PPP1R147, hSpred1, spred-1
Location
Chr.15:38252087-38357249 on build GRCh38
Assay Gene Location
Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
SPRED1 NM_152594.2 NP_689807.1
XM_005254202.3 XP_005254259.1
XM_011521289.2 XP_011519591.1
AK091222.1 BAC03614.1
AY299089.1 AAP59414.1
BC018015.1
BC137480.1
BC137481.1
DA405650.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv3892674 Chr15:37311235 - 38400599 on Build GRCh38 Gain SPRED1 LOC101928227 TMCO5A
nsv832974 Chr15:38206405 - 38398384 on Build GRCh38 Loss SPRED1
nsv832973 Chr15:38151604 - 38352681 on Build GRCh38 Gain SPRED1

View Full Product Details