Product Details
- Assay Reference Genome
Location
Chr.21:25885270 on build GRCh38
- Cytoband
- 21q21.3
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
For this assay, SNP(s) [rs116163905] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Target Gene Details
- Entrez Gene ID
- 351
- Gene Symbol
- APP
- Gene Name
- amyloid beta precursor protein
- Gene Aliases
- AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN-II, PN2
- Location
- Chr.21:25880550-26171128 on build GRCh38
- Assay Gene Location
- Within Intron 19
View Full Product Details