Product Details
- Assay Reference Genome
Location
Chr.21:25888634 on build GRCh38
- Cytoband
- 21q21.3
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 351
- Gene Symbol
- APP
- Gene Name
- amyloid beta precursor protein
- Gene Aliases
- AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN-II, PN2
- Location
- Chr.21:25880550-26171128 on build GRCh38
- Assay Gene Location
- Within Intron 19
View Full Product Details