Product Details
- Assay Reference Genome
Location
Chr.X:9729325 on build GRCh38
- Cytoband
- Xp22.2
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 4935
- Gene Symbol
- GPR143
- Gene Name
- G protein-coupled receptor 143
- Gene Aliases
- NYS6, OA1
- Location
- Chr.X:9725413-9765965 on build GRCh38
- Assay Gene Location
- Within Intron 8
View Full Product Details