Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
PINK1 | NM_032409.2 | NP_115785.1 | ||
AB053323.1 | BAB55647.1 | |||
AF316873.1 | AAK28062.1 | |||
AK130746.1 | ||||
BC028215.1 | AAH28215.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv545731 | Chr1:19989106 - 20776306 on Build GRCh38 | Gain | MUL1 CDA UBXN10-AS1 VWA5B1 MIR6084 PLA2G2C DDOST HP1BP3 UBXN10 FAM43B SH2D5 PLA2G2D PLA2G5 PLA2G2F PINK1-AS LINC01141 CAMK2N1 KIF17 PINK1 |
nsv834369 | Chr1:20609770 - 20779837 on Build GRCh38 | Loss | SH2D5 CDA MIR6084 PINK1-AS DDOST KIF17 HP1BP3 PINK1 |