Product Details

Assay Reference Genome
Location

Chr.8:31057652 on build GRCh38
Cytoband
8p12
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
7486
Gene Symbol
WRN
Gene Name
Werner syndrome RecQ like helicase
Gene Aliases
RECQ3, RECQL2, RECQL3
Location
Chr.8:31033262-31173761 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
WRN NM_000553.4 NP_000544.2
XM_011544639.2 XP_011542941.1
AF091214.1 AAC63361.1
BP279291.1

View Full Product Details