Product Details

Assay Reference Genome
Location

Chr.9:77246667 on build GRCh38
Cytoband
9q21.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
23230
Gene Symbol
VPS13A
Gene Name
vacuolar protein sorting 13 homolog A
Gene Aliases
CHAC, CHOREIN
Location
Chr.9:77177353-77421537 on build GRCh38
Assay Gene Location
Within Intron 19
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
VPS13A NM_001018037.1 NP_001018047.1
NM_001018038.2 NP_001018048.1
NM_015186.3 NP_056001.1
NM_033305.2 NP_150648.2
AB054005.1 BAB59128.1
AF337532.1 AAK61861.1
AJ608769.1 CAE75581.1
AJ626859.1 CAF25186.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1041647 Chr9:77211923 - 77537136 on Build GRCh38 Gain VPS13A GNA14 GNA14-AS1
nsv614728 Chr9:77195973 - 77375037 on Build GRCh38 Loss VPS13A

View Full Product Details