Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
AP1M1 | NM_001130524.1 | NP_001123996.1 | ||
NM_032493.3 | NP_115882.1 | |||
XM_017027420.1 | XP_016882909.1 | |||
XM_017027421.1 | XP_016882910.1 | |||
AB209808.1 | BAD93045.1 | |||
AF290613.1 | AAK28024.1 | |||
AK027528.1 | ||||
AK293188.1 | ||||
AK297824.1 | ||||
BC017469.2 | AAH17469.1 | |||
DQ059565.1 | AAY54246.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833767 | Chr19:16121391 - 16347760 on Build GRCh38 | Loss | FAM32A AP1M1 CIB3 HSH2D RAB8A KLF2 |