Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCL2
Entrez Gene ID
596
Gene Name
BCL2, apoptosis regulator
Gene Aliases
Bcl-2, PPP1R50
Chromosome Location
Chr.18: 63123346 - 63319778 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000633.2 NP_000624.2 2 - 3 1079 76
GenBank mRNA BC027258.1 - 2 - 3 975 76
EU287875.1 - 1 - 2 586 76
M13994.1 - 1 - 2 2044 76
M14745.1 - 1 - 2 617 76
X06487.1 - 2 - 3 1472 76

View Full Product Details