The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005508.4 | NP_005499.1 | 2 - 2 | 1194 | 70 |
XM_017005687.1 | XP_016861176.1 | 2 - 2 | 1241 | 70 | |
GenBank mRNA | AK292674.1 | - | 2 - 2 | 1124 | 70 |
BC069139.1 | - | 1 - 1 | 1079 | 70 | |
BC071751.1 | - | 2 - 2 | 1194 | 70 | |
BC074935.2 | - | 1 - 1 | 1054 | 70 |