Gene Transcripts

Gene Symbol
Pafah2
Entrez Gene ID
100163
Gene Name
platelet-activating factor acetylhydrolase 2
Gene Aliases
2310074E22Rik, AI507170
Chromosome Location
Chr. 4: 134396320 - 134427412 on Build GRCm38
UniGene ID
Mm.22116
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001285872.1 NP_001272801.1 9 1028
NM_001285874.1 NP_001272803.1 8 970
NM_001285875.1 NP_001272804.1 8 920
NM_001285877.1 NP_001272806.1 6 664
NM_133880.3 NP_598641.2 9 1017
GenBank mRNA AK144090.1 - 9 980
AK145092.1 - 8 871
AK156774.1 - 8 971
AK187928.1 - 31
AK205204.1 - 1 31
BC021890.1 - 8 922 3598000
BC025495.1 - 9 1039 5310200
BC025938.1 - 8 863

View Full Product Details