Gene Transcripts

Gene Symbol
Osbpl9
Entrez Gene ID
100273
Gene Name
oxysterol binding protein-like 9
Gene Aliases
2600011I06Rik, AU015843, Orp-9
Chromosome Location
Chr. 4: 109061145 - 109202272 on Build GRCm38
UniGene ID
Mm.366315
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134791.2 NP_001128263.1 16 1555
NM_133885.4 NP_598646.4 19 1788
NM_173350.3 NP_775485.2 17 1594
GenBank mRNA AK035658.1 - 17 1543
AK075981.1 - 6 573
AK131984.1 - 19 1759
AK172010.1 - 17 1581
AK178534.1 - 2 104
AK202959.1 - 2 104
AK207117.1 - 2 104
BC021507.1 - 15 1416 5347121
BC023759.1 - 19 1743
BC026927.1 - 17 1477 5134780
BC089163.1 - 16 1496 5367603

View Full Product Details