Gene Transcripts
- Gene Symbol
- Exoc5
- Entrez Gene ID
- 105504
- Gene Name
- exocyst complex component 5
- Gene Aliases
- AI447711, AI448003, Gm76, PRO1912, SEC10, Sec10l1
- Chromosome Location
- Chr. 14: 49011922 - 49066667 on Build GRCm38
- UniGene ID
- Mm.31607
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_207214.3 |
NP_997097.1
|
17, 18 |
2450 |
|
GenBank mRNA |
AK134562.1 |
- |
17, 18 |
1963 |
|
AK150297.1 |
- |
6, 7 |
717 |
|
AK152307.1 |
- |
17, 18 |
2179 |
|
AK157473.1 |
- |
17, 18 |
2145 |
|
AK164067.1 |
- |
17, 18 |
2172 |
|
AK167145.1 |
- |
17, 18 |
2138 |
|
AK170148.1 |
- |
17, 18 |
2144 |
|
AK171012.1 |
- |
17, 18 |
2168 |
|
BC049967.1 |
- |
17, 18 |
2107 |
3672313 |
View Full Product Details