Gene Transcripts

Gene Symbol
Slc35b3
Entrez Gene ID
108652
Gene Name
solute carrier family 35, member B3
Gene Aliases
4921526O06Rik, AI428480, CGI-19, PABST2, PAPST2
Chromosome Location
Chr. 13: 38932140 - 38960875 on Build GRCm38
UniGene ID
Mm.255506
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170430.1 NP_001163901.1 3 497
NM_001170431.1 NP_001163902.1 3 632
NM_134060.4 NP_598821.1 3 475
GenBank mRNA AK076592.1 - 3 470
AK077208.1 - 3 451
AK141280.1 - 3 633
BB851850.1 - 3 472
BC006881.1 - 3 489 3596363

View Full Product Details