Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289730.1 | NP_001276659.1 | 5 | 710 | |
NM_001289731.1 | NP_001276660.1 | 8 | 1266 | ||
NM_133197.4 | NP_573460.2 | 5 | 710 | ||
GenBank mRNA | AB052945.1 | - | 5 | 653 | |
AB101616.1 | - | 5 | 653 | ||
AK032122.1 | - | 5 | 711 | ||
AK045424.1 | - | 8 | 1268 | ||
BC125435.1 | - | 5 | 612 | 40129950 |