Gene Transcripts
- Gene Symbol
- Aebp1
- Entrez Gene ID
- 11568
- Gene Name
- AE binding protein 1
- Gene Aliases
- ACLP
- Chromosome Location
- Chr. 11: 5861853 - 5872225 on Build GRCm38
- UniGene ID
- Mm.3317
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF053943.1 |
- |
20, 21 |
2978 |
|
AK004712.1 |
- |
16, 17 |
1987 |
|
AK128980.1 |
- |
20, 21 |
3054 |
|
AK158276.1 |
- |
20, 21 |
3004 |
|
AK159330.1 |
- |
20, 21 |
2985 |
|
AK159342.1 |
- |
20, 21 |
3004 |
|
AK159377.1 |
- |
20, 21 |
2975 |
|
AK159409.1 |
- |
20, 21 |
3005 |
|
AK159774.1 |
- |
20, 21 |
2976 |
|
AK159957.1 |
- |
20, 21 |
2976 |
|
AK208974.1 |
- |
3, 4 |
286 |
|
AK210444.1 |
- |
3, 4 |
282 |
|
AK216593.1 |
- |
3, 4 |
281 |
|
BC082577.1 |
- |
20, 21 |
2990 |
30648530 |
X80478.1 |
- |
11, 12 |
1708 |
|
View Full Product Details