Gene Transcripts
- Gene Symbol
- Afp
- Entrez Gene ID
- 11576
- Gene Name
- alpha fetoprotein
- Gene Aliases
- Chromosome Location
- Chr. 5: 90490714 - 90508907 on Build GRCm38
- UniGene ID
- Mm.358570
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_007423.4 |
NP_031449.3
|
1, 2 |
149 |
|
GenBank mRNA |
AK010934.1 |
- |
1, 2 |
125 |
|
AK075972.1 |
- |
1, 2 |
129 |
|
AK076053.1 |
- |
1, 2 |
125 |
|
AK076197.1 |
- |
1, 2 |
127 |
|
AK145975.1 |
- |
1, 2 |
125 |
|
AK146513.1 |
- |
1, 2 |
127 |
|
AK146524.1 |
- |
1, 2 |
127 |
|
AK167549.1 |
- |
1, 2 |
125 |
|
AK168426.1 |
- |
1, 2 |
127 |
|
AK168574.1 |
- |
1, 2 |
127 |
|
AK168576.1 |
- |
1, 2 |
125 |
|
AK168751.1 |
- |
1, 2 |
127 |
|
AK168883.1 |
- |
1, 2 |
125 |
|
AK169046.1 |
- |
1, 2 |
127 |
|
AK169054.1 |
- |
1, 2 |
127 |
|
AK169147.1 |
- |
1, 2 |
125 |
|
BC066206.1 |
- |
1, 2 |
112 |
30469872 |
BY773400.1 |
- |
1, 2 |
156 |
|
V00743.1 |
- |
1, 2 |
123 |
|
View Full Product Details