Gene Transcripts

Gene Symbol
Bsn
Entrez Gene ID
12217
Gene Name
bassoon
Gene Aliases
Chromosome Location
Chr. 9: 108096022 - 108191938 on Build GRCm38
UniGene ID
Mm.20425
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007567.2 NP_031593.2 6 9815
GenBank mRNA AK129141.1 - 2 1375

View Full Product Details