Gene Transcripts

Gene Symbol
C3ar1
Entrez Gene ID
12267
Gene Name
complement component 3a receptor 1
Gene Aliases
AZ3B, C3AR, HNFAG09
Chromosome Location
Chr. 6: 122847140 - 122856157 on Build GRCm38
UniGene ID
Mm.2408
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009779.2 NP_033909.1 2 542
GenBank mRNA AK054017.1 - 2 545
AK154134.1 - 2 468
AK154229.1 - 2 530
AK154609.1 - 2 469
AY786554.1 - 1 360
BB665738.1 - 2 542
BC003728.1 - 2 455 3593758
U77460.1 - 2 390

View Full Product Details