Gene Transcripts
- Gene Symbol
- Aspm
- Entrez Gene ID
- 12316
- Gene Name
- asp (abnormal spindle)-like, microcephaly associated (Drosophila)
- Gene Aliases
- Calmbp1, D330028K02Rik, MCPH5, Sha1
- Chromosome Location
- Chr. 1: 139454773 - 139494091 on Build GRCm38
- UniGene ID
- Mm.168523
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009791.4 |
NP_033921.3
|
18 |
6431 |
|
GenBank mRNA |
AF533752.1 |
- |
18 |
6409 |
|
View Full Product Details