Gene Transcripts

Gene Symbol
Cat
Entrez Gene ID
12359
Gene Name
catalase
Gene Aliases
2210418N07, Cas-1, Cas1, Cs-1
Chromosome Location
Chr. 2: 103453904 - 103485153 on Build GRCm38
UniGene ID
Mm.4215
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009804.2 NP_033934.2 7 897
GenBank mRNA AK075853.1 - 7 867
AK088168.1 - 7 859
AK128919.1 - 7 872
AK133881.1 - 7 865
AK148961.1 - 7 865
AK150893.1 - 7 868
AK159152.1 - 7 868
AK159885.1 - 7 868
AK159891.1 - 7 819
AK159913.1 - 7 849
AK169069.1 - 7 862
AY040626.1 - 7 838
BC013447.1 - 7 854 4241333
BC047126.1 - 7 812
L25069.1 - 7 869
M62897.1 - 7 782
X52108.1 - 7 782

View Full Product Details