Gene Transcripts

Gene Symbol
Cr2
Entrez Gene ID
12902
Gene Name
complement receptor 2
Gene Aliases
C3DR, CD21, CD35, Cr-1, Cr-2, Cr1
Chromosome Location
Chr. 1: 195136811 - 195177190 on Build GRCm38
UniGene ID
Mm.235387
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007758.2 NP_031784.1 3 606
GenBank mRNA AK003072.2 - 3 606
BC109344.1 - 3 539 40054528
M35684.1 - 2 474
M61132.1 - 2 441

View Full Product Details