Gene Transcripts

Gene Symbol
Dll1
Entrez Gene ID
13388
Gene Name
delta-like 1 (Drosophila)
Gene Aliases
Delta1
Chromosome Location
Chr. 17: 15367354 - 15376113 on Build GRCm38
UniGene ID
Mm.4875
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007865.3 NP_031891.2 10 2663
GenBank mRNA BC057400.1 - 10 2176
BC065063.1 - 10 2427 6416191
X80903.1 - 10 2093

View Full Product Details