Gene Transcripts

Gene Symbol
Dsc3
Entrez Gene ID
13507
Gene Name
desmocollin 3
Gene Aliases
5430426I24Rik
Chromosome Location
Chr. 18: 19960930 - 20002413 on Build GRCm38
UniGene ID
Mm.89935
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291809.1 NP_001278738.1 13 2219
NM_007882.3 NP_031908.3 13 2219
GenBank mRNA BC144845.1 - 13 2204
Y11169.1 - 13 2219

View Full Product Details