Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010111.5 | NP_034241.2 | 5 | 837 | |
GenBank mRNA | AK132512.1 | - | 5 | 1304 | |
AK132845.1 | - | 5 | 1130 | ||
AK134108.1 | - | 5 | 1304 | ||
AK142203.1 | - | 5 | 1307 | ||
AK142283.1 | - | 5 | 1307 | ||
AK143059.1 | - | 5 | 1304 | ||
BC057009.1 | - | 5 | 1125 | 6827408 | |
CT010381.1 | - | 5 | 678 | ||
L38847.1 | - | 5 | 838 | ||
U16819.1 | - | 5 | 706 | ||
U30244.1 | - | 5 | 675 |