Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_080643.2 | NP_542374.2 | 15 | 1966 | |
GenBank mRNA | AF451978.1 | - | 14 | 1685 | |
AK037133.1 | - | 2 | 91 | ||
AK129297.1 | - | 15 | 2004 | ||
BC053083.1 | - | 15 | 1966 | 5704642 |