Gene Transcripts

Gene Symbol
Il1r2
Entrez Gene ID
16178
Gene Name
interleukin 1 receptor, type II
Gene Aliases
CD121b, Il1r-2
Chromosome Location
Chr. 1: 40081750 - 40125230 on Build GRCm38
UniGene ID
Mm.1349
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010555.4 NP_034685.1 8 1078
GenBank mRNA AK132264.1 - 8 1105
BC032962.1 - 7 795 1282531
CT010182.1 - 7 986
X59769.1 - 8 1078

View Full Product Details